This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01296202_g1
See other MAP3K12 GE Assays ›
Gene Symbol:
Map3k12
Gene Name:
mitogen-activated protein kinase kinase kinase 12
Gene Aliases:
DLK, MUK, Zpk
Chromosome Location:
Chr.15: 102497647 - 102518053 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01296202_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Map3k12
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163643.1 NP_001157115.1 13 - 14 2662 122
NM_009582.4 NP_033608.3 11 - 12 2486 122
GenBank mRNA AK029882.1 - 11 - 12 2295 122
AK034374.1 - 13 - 14 2478 122
AK081623.1 - 6 - 7 1154 122
AK166435.1 - 13 - 14 2582 122
BC047158.1 - 10 - 11 2280 6494216 122
BC057572.1 - 13 - 14 2486 6401321 122
U14636.1 - 11 - 12 2318 122
U23789.1 - 10 - 11 2276 122

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500002294.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Map3k12

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

activation of MAPKK activity
protein phosphorylation
JNK cascade
phosphorylation
histone phosphorylation
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
protein autophosphorylation
mitophagy in response to mitochondrial depolarization
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
MAP kinase kinase kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
protein kinase binding
protein homodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500018957.mm.1 NM_001163643.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500018957.mm.1 NM_001163643.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500018957.mm.1 NM_009582.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500018957.mm.1 NM_009582.4 Refseq
Affymetrix Mouse Clariom S Array PSR1500018957.mm.1 TC1500002294.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0