This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01295883_m1
See other TNR GE Assays ›
Gene Symbol:
Tnr
Gene Name:
tenascin R
Gene Aliases:
J1-tenascin, TN-R, janusin, restrictin
Chromosome Location:
Chr.1: 159523741 - 159931729 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01295883_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tnr
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_022312.3 NP_071707.2 21 - 22 4402 64
GenBank mRNA AK039390.1 - 21 - 22 4403 64
AK078229.1 - 14 - 15 2080 64
BC132392.1 - 21 - 22 4036 40130669 64
BC138043.1 - 21 - 22 4036 8861062 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100001447.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tnr

Panther Classification:

Molecular Function -

extracellular matrix protein

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
neuron cell-cell adhesion
negative regulation of cell adhesion
associative learning
negative regulation of neuron projection development
telencephalon cell migration
negative regulation of cell-cell adhesion
extracellular matrix organization
negative regulation of axon extension
locomotory exploration behavior
negative regulation of axon extension involved in regeneration
modulation of synaptic transmission
negative regulation of synaptic transmission
synapse organization
neuromuscular process controlling balance
positive regulation of synaptic transmission, glutamatergic
positive regulation of transmission of nerve impulse
long-term synaptic potentiation
integrin binding
sphingolipid binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100011865.mm.1 NM_022312.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100011865.mm.1 NM_022312.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100011865.mm.1 TC0100001447.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0