This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01293020_m1
See other PRKAR2B GE Assays ›
Gene Symbol:
Prkar2b
Gene Name:
protein kinase, cAMP dependent regulatory, type II beta
Gene Aliases:
AI451071, AW061005, PKARIIbeta, Pkarb2, RII(beta)
Chromosome Location:
Chr.12: 31958450 - 32061343 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01293020_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Prkar2b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011158.3 NP_035288.2 7 - 8 1030 122
GenBank mRNA AK011647.1 - 7 - 8 988 122
AK041013.1 - 7 - 8 1004 122
AK081805.1 - 7 - 8 807 122
AK135621.1 - 7 - 8 1030 122
AK138963.1 - 7 - 8 996 122
BC033267.1 - 3 - 4 318 122
BC048710.1 - 7 - 8 986 6506827 122

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Prkar2b

Panther Classification:

Molecular Function -

protein-binding activity modulator kinase modulator

Gene Ontology Categories:

Function(s) Process(es)

regulation of protein phosphorylation
fatty acid metabolic process
learning
regulation of protein kinase activity
negative regulation of cAMP-dependent protein kinase activity
nucleotide binding
cAMP-dependent protein kinase inhibitor activity
protein binding
cAMP-dependent protein kinase regulator activity
protein kinase binding
protein domain specific binding
cAMP binding
ubiquitin protein ligase binding
protein kinase A catalytic subunit binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0