This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01292960_g1
See other GEMIN2 GE Assays ›
Gene Symbol:
Gemin2
Gene Name:
gem (nuclear organelle) associated protein 2
Gene Aliases:
1700012N19Rik, Sip1
Chromosome Location:
Chr.12: 59013393 - 59027988 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01292960_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gemin2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025656.4 NP_079932.2 7 - 8 684 153
GenBank mRNA AK005928.1 - 7 - 8 686 153
AK007515.1 - 7 - 8 685 153
AK013414.1 - 7 - 8 641 153
BC053424.1 - 7 - 8 1113 6336522 153

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gemin2

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s)

spliceosomal complex assembly
spliceosomal snRNP assembly
mRNA splicing, via spliceosome
mRNA processing
RNA splicing

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200003458.mm.1 NM_025656.5 Refseq
Affymetrix Mouse Clariom S Array PSR1200003458.mm.1 TC1200000498.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0