This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01292931_g1
See other CTAGE5 GE Assays ›
Gene Symbol:
Ctage5
Gene Name:
CTAGE family, member 5
Gene Aliases:
D12Bwg0579e, Mea6, Mgea, Mgea6
Chromosome Location:
Chr.12: 59129740 - 59190220 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01292931_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ctage5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001165253.1 NP_001158725.1 3 - 4 307 71
NM_001165254.1 NP_001158726.1 3 - 4 306 71
NM_146034.3 NP_666146.3 3 - 4 306 71
GenBank mRNA AK029466.1 - 3 - 4 306 71
AK053010.1 - 3 - 4 240 71
AK080312.1 - 3 - 4 282 71
AK186274.1 - 2 - 3 100 71
BC026864.1 - 3 - 4 258 2647214 71
BQ571563.1 - 3 - 4 306 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1200000503.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ctage5

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200003509.mm.1 NM_001165253.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200003509.mm.1 NM_001165253.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200003509.mm.1 NM_001165254.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200003509.mm.1 NM_001165254.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200003509.mm.1 NM_146034.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200003509.mm.1 NM_146034.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200003509.mm.1 TC1200000503.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200003505.mm.1 NM_001165253.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200003505.mm.1 NM_001165253.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200003505.mm.1 NM_001165254.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200003505.mm.1 NM_001165254.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200003505.mm.1 NM_146034.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200003505.mm.1 NM_146034.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200003505.mm.1 TC1200000503.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200003506.mm.1 NM_146034.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200003506.mm.1 NM_146034.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200003506.mm.1 TC1200000503.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0