This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01292926_g1
See other CTAGE5 GE Assays ›
Gene Symbol:
Ctage5
Gene Name:
CTAGE family, member 5
Gene Aliases:
D12Bwg0579e, Mea6, Mgea, Mgea6
Chromosome Location:
Chr.12: 59129740 - 59190220 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01292926_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ctage5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001165253.1 NP_001158725.1 14 - 15 1357 86
NM_001165254.1 NP_001158726.1 14 - 15 1356 86
NM_146034.3 NP_666146.3 16 - 17 1455 86
GenBank mRNA AK029466.1 - 14 - 15 1356 86
AK138989.1 - 2 - 3 2303 86
AK157358.1 - 14 - 15 1358 86
BC018567.1 - 8 - 9 734 86
BC024076.1 - 7 - 8 695 86
BC026864.1 - 16 - 17 1413 2647214 86

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ctage5

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200003528.mm.1 NM_001165253.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200003528.mm.1 NM_001165253.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200003528.mm.1 NM_001165254.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200003528.mm.1 NM_001165254.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200003528.mm.1 NM_146034.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200003528.mm.1 NM_146034.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200003528.mm.1 TC1200000503.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200003530.mm.1 NM_001165253.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200003530.mm.1 NM_001165253.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200003530.mm.1 NM_001165254.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200003530.mm.1 NM_001165254.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200003530.mm.1 NM_146034.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200003530.mm.1 NM_146034.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200003530.mm.1 TC1200000503.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200003531.mm.1 NM_001165253.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200003531.mm.1 NM_001165253.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200003531.mm.1 NM_001165254.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200003531.mm.1 NM_001165254.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200003531.mm.1 NM_146034.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200003531.mm.1 NM_146034.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200003531.mm.1 TC1200000503.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0