This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01292107_m1
See other DARS GE Assays ›
Gene Symbol:
Dars
Gene Name:
aspartyl-tRNA synthetase
Gene Aliases:
5730439G15Rik
Chromosome Location:
Chr.1: 128363707 - 128417416 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01292107_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dars
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_177445.5 NP_803228.2 6 - 7 700 91
GenBank mRNA AK077533.1 - 6 - 7 623 91
AK144470.1 - 6 - 7 645 91
AK146350.1 - 6 - 7 644 91
AK151027.1 - 6 - 7 644 91
AK151897.1 - 6 - 7 611 91
AK152304.1 - 6 - 7 611 91
AK152562.1 - 6 - 7 611 91
AK153256.1 - 6 - 7 611 91
AK153299.1 - 6 - 7 611 91
AK166009.1 - 6 - 7 611 91
AK169247.1 - 6 - 7 611 91
AK169716.1 - 6 - 7 588 91
AK193968.1 - 2 - 3 157 91
BC008638.1 - 6 - 7 598 3586278 91

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dars

Panther Classification:

Molecular Function -

aminoacyl-tRNA synthetase

Gene Ontology Categories:

Function(s) Process(es)

translation
tRNA aminoacylation for protein translation
aspartyl-tRNA aminoacylation
nucleotide binding
nucleic acid binding
aminoacyl-tRNA ligase activity
aspartate-tRNA ligase activity
ATP binding
ligase activity
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0