This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01291803_m1
See other AGR2 GE Assays ›
Gene Symbol:
Agr2
Gene Name:
anterior gradient 2
Gene Aliases:
Agr2h, Gob-4, HAG-2, XAG-2, mAG-2
Chromosome Location:
Chr.12: 35992925 - 36004081 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01291803_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Agr2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011783.2 NP_035913.1 5 - 6 386 70
GenBank mRNA AK007677.1 - 5 - 6 405 70
AK008296.1 - 4 - 5 454 70

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Agr2

Panther Classification:

Molecular Function -

reductase

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of gene expression
positive regulation of cell-substrate adhesion
positive regulation of epidermal growth factor receptor signaling pathway
lung goblet cell differentiation
negative regulation of cell death
mucus secretion
positive regulation of establishment of protein localization to plasma membrane
positive regulation of IRE1-mediated unfolded protein response
positive regulation of PERK-mediated unfolded protein response
dystroglycan binding
epidermal growth factor receptor binding
protein homodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0