Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200011113.mm.1 |
NM_175191.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011113.mm.1 |
NM_175191.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011113.mm.1 |
TC1200001599.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200011110.mm.1 |
NM_175191.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011110.mm.1 |
NM_175191.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011110.mm.1 |
TC1200001599.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200011111.mm.1 |
NM_175191.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011111.mm.1 |
NM_175191.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011111.mm.1 |
TC1200001599.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200011112.mm.1 |
NM_175191.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011112.mm.1 |
NM_175191.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011112.mm.1 |
TC1200001599.mm.1 |
MTA TC |