This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01290843_g1
See other ATP1A4 GE Assays ›
Gene Symbol:
Atp1a4
Gene Name:
ATPase, Na+/K+ transporting, alpha 4 polypeptide
Gene Aliases:
-
Chromosome Location:
Chr.1: 172223508 - 172258424 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01290843_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atp1a4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013734.1 NP_038762.1 12 - 13 132

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100003565.mm.1

Set Membership:

Amplicon length greater than or equal to 101 Ion Channel Transporter Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

primary active transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
potassium ion transport
sodium ion transport
spermatogenesis
fertilization
sperm motility
regulation of cellular pH
sodium ion transmembrane transport
regulation of membrane potential
potassium ion transmembrane transport
nucleotide binding
sodium:potassium-exchanging ATPase activity
ATP binding
hydrolase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100029279.mm.1 NM_013734.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100029279.mm.1 NM_013734.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100029279.mm.1 TC0100003565.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0