Assay Details
Gene Symbol: Nxpe5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013773.3 | NP_001013795.3 | 3 - 4 | 1252 | 99 | |
GenBank mRNA | AK156114.1 | - | 3 - 4 | 1254 | 99 | |
BC055004.1 | - | 868 | 6478735 | 99 | ||
BC106195.1 | - | 1050 | 6396133 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24129471 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24129473 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24129472 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24129474 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2026139 | NM_001013773.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2026139 | AK156114.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 692513 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 898765 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 447489 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 776517 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 46801 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 767924 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 854956 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 736626 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 305672 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 207589 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 882076 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 366137 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 13219 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 102099 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 153592 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P130823 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2168511 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2026139 | NM_001013773.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2026139 | AK156114.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24129389 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24129402 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24129396 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24129381 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24129398 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24129386 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24129405 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24129383 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24129391 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24129385 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24129377 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24129408 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24129394 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24129393 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24129379 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24129397 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24129401 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24129378 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24129406 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24129403 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01232760 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05762527 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05762555 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P130823 | - | - |
Whole Mouse Genome G4122A | A_52_P136348 | BC106195.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P136348 | BC055004.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01232760 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P130823 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04772262 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04772255 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04772258 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24129472 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24129471 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24129473 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24129474 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129497 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129464 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129474 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129453 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129406 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129377 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129401 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129397 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129480 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129425 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129405 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129435 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129456 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129459 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129501 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129394 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129492 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129447 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129514 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129440 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129408 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129508 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129398 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129450 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129472 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129424 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129516 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129462 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129476 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129519 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129427 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129477 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129471 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129506 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129433 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129466 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129483 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129370 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129505 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129439 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129510 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129487 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129494 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129402 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129479 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129437 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129441 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129378 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129457 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129452 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129461 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129470 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129391 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129379 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129451 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129434 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129475 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129389 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129399 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129513 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129495 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129499 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129383 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04747151 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129448 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129458 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04514042 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129387 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04514043 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129385 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129373 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129377 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04815947 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129394 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04815948 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04514045 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04530839 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129399 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129384 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04727427 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129389 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04884731 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04196200 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129451 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129391 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129375 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04192696 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129450 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129397 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129365 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129402 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129408 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04924158 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129367 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04772262 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129405 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04628097 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129398 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129406 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129381 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04367338 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04650722 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129370 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10526784 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10526788 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10526785 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10526791 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10526789 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10526787 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10526786 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10526790 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10526783 | - | - |
Affymetrix Mouse Clariom S Array | PSR0500015125.mm.1 | NM_001013773.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500015125.mm.1 | NM_001013773.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500015125.mm.1 | TC0500001648.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500015132.mm.1 | TC0500001648.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500015121.mm.1 | NM_001013773.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500015121.mm.1 | NM_001013773.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500015121.mm.1 | TC0500001648.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500015127.mm.1 | TC0500001648.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500015122.mm.1 | NM_001013773.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500015122.mm.1 | NM_001013773.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500015122.mm.1 | TC0500001648.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500015124.mm.1 | NM_001013773.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500015124.mm.1 | NM_001013773.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500015124.mm.1 | TC0500001648.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 411009 | - | - |
ABI Mouse Genome Survey Microarray | 714772 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.1 expression beadchip | ILMN_1258189 | - | - |
