This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01290782_m1
See other D630008O14RIK GE Assays ›
Gene Symbol:
D630008O14Rik
Gene Name:
RIKEN cDNA D630008O14 gene
Gene Aliases:
D630044D05Rik
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01290782_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: D630008O14Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK085305.1 - 2 - 3 554 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1973593 AK085305.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1973593 AK085305.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123872 - -
Array Name Probe IDs Targets Target Type
Mouse Genome 430 2.0 Array 1437122_at - -
Mouse Genome 430 2.0 Array 1437122_PM_at - -
Mouse Expression Set 430 1437122_at - -
Mouse Expression Set 430 1437122_PM_at - -
Murine Genome U74v2 Set 137975_i_at - -
Murine Genome U74v2 Set 137975_PM_i_at - -
Murine Genome U74v2 Set 137976_r_at - -
Murine Genome U74v2 Set 137976_PM_r_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 468145 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 101500301 - -
Illumina mouse-6 v1.0 expression beadchip 101500301 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1251112 - -
Illumina Sentrix mouse-6.v1 bead array 101500301 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 101500301 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1251112 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0