This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01290710_g1
See other TOMM40L GE Assays ›
Gene Symbol:
Tomm40l
Gene Name:
translocase of outer mitochondrial membrane 40 homolog-like (yeast)
Gene Aliases:
-
Chromosome Location:
Chr.1: 171217802 - 171222514 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01290710_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tomm40l
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001037170.2 NP_001032247.1 8 - 9 961 93
GenBank mRNA AK012264.1 - 8 - 9 891 93
AK028388.1 - 8 - 9 962 93
AK029485.1 - 5 - 6 457 93
AK208323.1 - 1 - 2 36 93
BC058978.1 - 7 - 8 852 5715769 93

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tomm40l

Panther Classification:

Molecular Function -

primary active transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
biological_process
protein transport
transmembrane transport
molecular_function
porin activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100028881.mm.1 NM_001037170.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100028881.mm.1 NM_001037170.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100028881.mm.1 TC0100003532.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100028882.mm.1 NM_001037170.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100028882.mm.1 NM_001037170.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100028882.mm.1 TC0100003532.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100028883.mm.1 NM_001037170.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100028883.mm.1 NM_001037170.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100028883.mm.1 TC0100003532.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100028880.mm.1 NM_001037170.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100028880.mm.1 NM_001037170.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100028880.mm.1 TC0100003532.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0