Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100022194.mm.1 |
NM_008801.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100022194.mm.1 |
NM_008801.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100022194.mm.1 |
TC0100002748.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100022193.mm.1 |
NM_008801.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100022193.mm.1 |
NM_008801.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100022193.mm.1 |
TC0100002748.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100022197.mm.1 |
NM_008801.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100022197.mm.1 |
NM_008801.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100022197.mm.1 |
TC0100002748.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100022195.mm.1 |
NM_008801.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100022195.mm.1 |
NM_008801.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100022195.mm.1 |
TC0100002748.mm.1 |
MTA TC |