This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01290396_m1
See other CAB39 GE Assays ›
Gene Symbol:
Cab39
Gene Name:
calcium binding protein 39
Gene Aliases:
AA408805, AA960512, C78372, MO25, MO25alpha
Chromosome Location:
Chr.1: 85793410 - 85851577 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01290396_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cab39
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_133781.4 NP_598542.3 1 - 2 375 74
GenBank mRNA AK144665.1 - 2 - 3 208 74
CJ162690.1 - 1 - 2 377 74

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK144665.1, NM_133781.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cab39

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of peptidyl-threonine phosphorylation
signal transduction by protein phosphorylation
activation of protein kinase activity
positive regulation of peptidyl-serine phosphorylation
intracellular signal transduction
positive regulation of protein serine/threonine kinase activity
calcium ion binding
kinase binding
protein kinase activator activity
protein serine/threonine kinase activator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100006419.mm.1 NM_133781.5 Refseq
Affymetrix Mouse Clariom S Array PSR0100006419.mm.1 TC0100000771.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100006425.mm.1 NM_133781.5 Refseq
Affymetrix Mouse Clariom S Array PSR0100006425.mm.1 TC0100000771.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0