This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01290133_m1
See other HAPLN1 GE Assays ›
Gene Symbol:
Hapln1
Gene Name:
hyaluronan and proteoglycan link protein 1
Gene Aliases:
BB099155, CLP, Crtl1, Crtl1l, LP, LP-1
Chromosome Location:
Chr.13: 89540529 - 89611832 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01290133_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hapln1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013500.4 NP_038528.3 1 - 2 273 110
GenBank mRNA AF139572.1 - 1081 110
AK032750.1 - 1 - 2 274 110

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF139572.1, AK032750.1, NM_013500.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hapln1

Panther Classification:

Molecular Function -

extracellular matrix glycoprotein

Gene Ontology Categories:

Function(s) Process(es)

skeletal system development
cell adhesion
central nervous system development
mitophagy in response to mitochondrial depolarization
hyaluronic acid binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300006441.mm.1 NM_013500.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006441.mm.1 NM_013500.4 Refseq
Affymetrix Mouse Clariom S Array PSR1300006441.mm.1 TC1300001006.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300006439.mm.1 NM_013500.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006439.mm.1 NM_013500.4 Refseq
Affymetrix Mouse Clariom S Array PSR1300006439.mm.1 TC1300001006.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300006440.mm.1 NM_013500.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006440.mm.1 NM_013500.4 Refseq
Affymetrix Mouse Clariom S Array PSR1300006440.mm.1 TC1300001006.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300006442.mm.1 NM_013500.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006442.mm.1 NM_013500.4 Refseq
Affymetrix Mouse Clariom S Array PSR1300006442.mm.1 TC1300001006.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0