This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01290115_g1
See other EPB4.1L5 GE Assays ›
Gene Symbol:
Epb4.1l5
Gene Name:
erythrocyte protein band 4.1 like 5
Gene Aliases:
1700030C16Rik, AL022914, BE37, E230025E14Rik, Epb41l5, Lulu1, NBL5
Chromosome Location:
Chr.1: 119545032 - 119649136 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01290115_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Epb4.1l5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001113416.1 NP_001106887.1 8 - 9 841 130
NM_145506.4 NP_663481.2 8 - 9 841 130
GenBank mRNA AK034676.1 - 8 - 9 763 130
AK037024.1 - 8 - 9 797 130
AK040354.1 - 8 - 9 809 130
AK044959.1 - 8 - 9 779 130
AK044996.1 - 8 - 9 787 130
AK047523.1 - 8 - 9 768 130
AK079301.1 - 8 - 9 792 130
AK136125.1 - 8 - 9 796 130
AK161141.1 - 8 - 9 792 130
AK167449.1 - 8 - 9 841 130
AK173198.1 - 8 - 9 775 130
BC003937.1 - 8 - 9 826 3486543 130
BC011476.1 - 8 - 9 775 4014009 130

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100002950.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Epb4.1l5

Gene Ontology Categories:

Function(s) Process(es)

cell morphogenesis involved in differentiation
in utero embryonic development
somitogenesis
epithelial to mesenchymal transition
neural plate morphogenesis
positive regulation of cell-matrix adhesion
epithelial cell morphogenesis
apical constriction
substrate-dependent cell migration, cell attachment to substrate
ectoderm development
endoderm development
mesoderm development
mesoderm migration involved in gastrulation
unidimensional cell growth
posttranscriptional regulation of gene expression
positive regulation of epithelial cell migration
positive regulation of epithelial to mesenchymal transition
negative regulation of cell-cell adhesion
actin cytoskeleton organization
actomyosin structure organization
negative regulation of protein binding
positive regulation of protein binding
somite rostral/caudal axis specification
axial mesoderm development
axial mesoderm morphogenesis
paraxial mesoderm development
embryonic foregut morphogenesis
positive regulation of focal adhesion assembly
regulation of establishment of protein localization
left/right axis specification
cellular response to transforming growth factor beta stimulus
protein binding
cytoskeletal protein binding
protein domain specific binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100023714.mm.1 NM_001113416.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100023714.mm.1 NM_001113416.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100023714.mm.1 NM_145506.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100023714.mm.1 NM_145506.4 Refseq
Affymetrix Mouse Clariom S Array PSR0100023714.mm.1 TC0100002950.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0