This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01289542_m1
See other EGFEM1 GE Assays ›
Gene Symbol:
Egfem1
Gene Name:
EGF-like and EMI domain containing 1
Gene Aliases:
6130401L20Rik, Efcab11
Chromosome Location:
Chr.3: 29080745 - 29691058 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01289542_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Egfem1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001167748.1 NP_001161220.1 7 - 8 1089 69
NM_029412.1 NP_083688.1 9 - 10 1260 69
GenBank mRNA AK018073.1 - 9 - 10 1335 69
AK032017.1 - 9 - 10 1104 69
BC117810.1 - 7 - 8 895 40092604 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300000186.mm.1

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

extracellular matrix protein

Gene Ontology Categories:

Function(s) Process(es)

biological_process
calcium ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300001013.mm.1 NM_001167748.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300001013.mm.1 NM_001167748.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300001013.mm.1 NM_029412.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300001013.mm.1 NM_029412.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300001013.mm.1 TC0300000186.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0