This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01289479_m1
See other HSPA4L GE Assays ›
Gene Symbol:
Hspa4l
Gene Name:
heat shock protein 4 like
Gene Aliases:
94kDa, AI461691, APG-1, Osp94
Chromosome Location:
Chr.3: 40744355 - 40796103 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01289479_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hspa4l
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011020.3 NP_035150.3 3 - 4 366 61
GenBank mRNA AB001926.1 - 3 - 4 332 61
AK032066.1 - 3 - 4 396 61
AK144225.1 - 3 - 4 403 61
AK163459.1 - 4 - 5 538 61
BC012712.1 - 3 - 4 435 4190161 61
BC057002.1 - 3 - 4 366 5702602 61
BC110662.1 - 3 - 4 386 4511154 61
D49482.1 - 3 - 4 360 61
U23921.1 - 3 - 4 483 61

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hspa4l

Gene Ontology Categories:

Function(s) Process(es)

protein folding
response to unfolded protein
nucleotide binding
protein binding
ATP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-fd98957d6-rbs6c:80/100.66.128.142:80.
git-commit: 278a916a254c4ca07f9ccbb2b9efa8cd7578b5be
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0