This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01288815_m1
See other CNOT6 GE Assays ›
Gene Symbol:
Cnot6
Gene Name:
CCR4-NOT transcription complex, subunit 6
Gene Aliases:
CCR4
Chromosome Location:
Chr.11: 49671497 - 49714004 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01288815_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cnot6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001290741.1 NP_001277670.1 6 - 7 721 136
NM_212484.2 NP_997649.1 7 - 8 1075 136
GenBank mRNA AK129307.1 - 1 - 2 2013 136
AY043269.1 - 6 - 7 719 136
BC023766.1 - 2 - 3 186 136
BC049984.1 - 2 - 3 245 136
BC057190.1 - 1 - 2 172 136
BC062950.1 - 7 - 8 1063 6840162 136
CF746424.1 - 5 - 6 555 136

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100002659.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cnot6

Panther Classification:

Molecular Function -

mRNA polyadenylation factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of translation
positive regulation of cell proliferation
positive regulation of cytoplasmic mRNA processing body assembly
gene silencing by RNA
exonucleolytic nuclear-transcribed mRNA catabolic process involved in deadenylation-dependent decay
nuclear-transcribed mRNA catabolic process, no-go decay
RNA phosphodiester bond hydrolysis, exonucleolytic
positive regulation of ligand-dependent nuclear receptor transcription coactivator activity
RNA binding
nuclease activity
exonuclease activity
exoribonuclease activity
poly(A)-specific ribonuclease activity
hydrolase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100024286.mm.1 NM_212484.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100024286.mm.1 TC1100002659.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0