This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01288551_m1
See other RAPGEF6 GE Assays ›
Gene Symbol:
Rapgef6
Gene Name:
Rap guanine nucleotide exchange factor (GEF) 6
Gene Aliases:
A530068K01, AI844632, BB085664, C030018K18Rik, PDZ-GEF2, Pdzgef2, RA-GEF-2, Ragef2, mKIAA4052
Chromosome Location:
Chr.11: 54521754 - 54699286 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01288551_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rapgef6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001252494.1 NP_001239423.1 1 - 2 277 91
NM_001252496.1 NP_001239425.1 1 - 2 277 91
NM_175258.4 NP_780467.2 1 - 2 277 91
GenBank mRNA AK045540.1 - 1 - 2 219 91
AK049228.1 - 1 - 2 233 91
AK167055.1 - 1 - 2 278 91
BC141182.1 - 1 - 2 176 9055794 91
BC145503.1 - 1 - 2 176 91

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100000629.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rapgef6

Panther Classification:

Molecular Function -

guanyl-nucleotide exchange factor

Gene Ontology Categories:

Function(s) Process(es)

microvillus assembly
positive regulation of GTPase activity
protein localization to plasma membrane
guanyl-nucleotide exchange factor activity
Ras GTPase binding
GTP-dependent protein binding
phosphatidic acid binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100005061.mm.1 NM_001252494.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100005061.mm.1 NM_001252494.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100005061.mm.1 NM_001252496.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100005061.mm.1 NM_001252496.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100005061.mm.1 NM_175258.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100005061.mm.1 NM_175258.4 Refseq
Affymetrix Mouse Clariom S Array PSR1100005061.mm.1 TC1100000629.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100005066.mm.1 NM_001252494.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100005066.mm.1 NM_001252494.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100005066.mm.1 NM_001252496.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100005066.mm.1 NM_001252496.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100005066.mm.1 NM_175258.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100005066.mm.1 NM_175258.4 Refseq
Affymetrix Mouse Clariom S Array PSR1100005066.mm.1 TC1100000629.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100005060.mm.1 NM_001252494.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100005060.mm.1 NM_001252494.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100005060.mm.1 NM_001252496.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100005060.mm.1 NM_001252496.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100005060.mm.1 NM_175258.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100005060.mm.1 NM_175258.4 Refseq
Affymetrix Mouse Clariom S Array PSR1100005060.mm.1 TC1100000629.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100005057.mm.1 NM_001252494.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100005057.mm.1 NM_001252494.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100005057.mm.1 NM_001252496.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100005057.mm.1 NM_001252496.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100005057.mm.1 NM_175258.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100005057.mm.1 NM_175258.4 Refseq
Affymetrix Mouse Clariom S Array PSR1100005057.mm.1 TC1100000629.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 491804 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0