Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100005088.mm.1 |
NM_001252498.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005088.mm.1 |
NM_001252498.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005088.mm.1 |
TC1100000629.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100005085.mm.1 |
NM_001252494.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005085.mm.1 |
NM_001252494.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005085.mm.1 |
NM_001252496.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005085.mm.1 |
NM_001252496.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005085.mm.1 |
NM_001252497.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005085.mm.1 |
NM_001252497.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005085.mm.1 |
NM_001252498.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005085.mm.1 |
NM_001252498.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005085.mm.1 |
NM_175258.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005085.mm.1 |
NM_175258.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005085.mm.1 |
TC1100000629.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100005087.mm.1 |
NM_001252494.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005087.mm.1 |
NM_001252494.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005087.mm.1 |
NM_001252496.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005087.mm.1 |
NM_001252496.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005087.mm.1 |
NM_001252497.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005087.mm.1 |
NM_001252497.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005087.mm.1 |
NM_001252498.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005087.mm.1 |
NM_001252498.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005087.mm.1 |
NM_175258.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005087.mm.1 |
NM_175258.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005087.mm.1 |
TC1100000629.mm.1 |
MTA TC |