This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01288446_m1
See other ETV1 GE Assays ›
Gene Symbol:
Etv1
Gene Name:
ets variant 1
Gene Aliases:
ER81, Etsrp81
Chromosome Location:
Chr.12: 38779656 - 38868215 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01288446_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Etv1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163154.1 NP_001156626.1 4 - 5 463 86
NM_007960.4 NP_031986.1 7 - 8 803 86
GenBank mRNA AK032224.1 - 4 - 5 465 86
AK035914.1 - 5 - 6 1194 86
AK144713.1 - 7 - 8 752 86
AW125172.1 - 1 - 2 86
BC005645.1 - 7 - 8 731 3257346 86
L10426.1 - 7 - 8 752 86

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Etv1

Panther Classification:

Molecular Function -

winged helix/forkhead transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
axon guidance
muscle organ development
mechanosensory behavior
cell differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200002550.mm.1 NM_001163154.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200002550.mm.1 NM_001163154.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200002550.mm.1 NM_007960.5 Refseq
Affymetrix Mouse Clariom S Array PSR1200002550.mm.1 TC1200000348.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200002554.mm.1 NM_001163154.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200002554.mm.1 NM_001163154.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200002554.mm.1 NM_007960.5 Refseq
Affymetrix Mouse Clariom S Array PSR1200002554.mm.1 TC1200000348.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200002552.mm.1 NM_001163154.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200002552.mm.1 NM_001163154.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200002552.mm.1 NM_007960.5 Refseq
Affymetrix Mouse Clariom S Array PSR1200002552.mm.1 TC1200000348.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200002553.mm.1 NM_001163154.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200002553.mm.1 NM_001163154.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200002553.mm.1 NM_007960.5 Refseq
Affymetrix Mouse Clariom S Array PSR1200002553.mm.1 TC1200000348.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0