This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01288350_m1
See other PEX2 GE Assays ›
Gene Symbol:
Pex2
Gene Name:
peroxisomal biogenesis factor 2
Gene Aliases:
D3Ertd138e, PAF-1, PMP35, Pxmp3
Chromosome Location:
Chr.3: 5560188 - 5576877 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01288350_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pex2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163305.2 NP_001156777.1 2 - 3 354 64
NM_008994.4 NP_033020.2 2 - 3 354 64
GenBank mRNA BC012404.1 - 2 - 3 234 3964491 64
CX231604.1 - 2 - 3 235 64
CX231749.1 - 2 - 3 185 64

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC012404.1, NM_001163305.2, NM_008994.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pex2

Gene Ontology Categories:

Function(s) Process(es)

very long-chain fatty acid metabolic process
negative regulation of transcription from RNA polymerase II promoter
protein polyubiquitination
neuron migration
fatty acid beta-oxidation
bile acid biosynthetic process
peroxisome organization
nervous system development
protein import into peroxisome matrix
protein destabilization
cholesterol homeostasis
regulation of cholesterol biosynthetic process
negative regulation of fibroblast proliferation
negative regulation of epithelial cell proliferation
ubiquitin-protein transferase activity
zinc ion binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300012841.mm.1 NM_001163301.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300012841.mm.1 NM_001163302.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300012841.mm.1 NM_001163305.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300012841.mm.1 NM_001163306.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300012841.mm.1 NM_001267714.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300012841.mm.1 NM_001267715.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300012841.mm.1 NM_008994.4 Refseq
Affymetrix Mouse Clariom S Array PSR0300012841.mm.1 TC0300001601.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0