This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01285640_m1
See other TRAF3IP1 GE Assays ›
Gene Symbol:
Traf3ip1
Gene Name:
TRAF3 interacting protein 1
Gene Aliases:
3930402D05Rik, AU041749, MIP-T3
Chromosome Location:
Chr.1: 91494590 - 91529307 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01285640_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Traf3ip1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028718.2 NP_082994.1 8 - 9 1175 64
GenBank mRNA AK014457.1 - 5 - 6 656 64
AY613437.1 - 8 - 9 1178 64
BC117868.2 - 8 - 9 1149 40095476 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100000897.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Traf3ip1

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
negative regulation of protein phosphorylation
neural tube patterning
embryonic camera-type eye development
negative regulation of protein complex assembly
negative regulation of type I interferon production
negative regulation of interferon-beta production
embryonic heart tube development
post-anal tail morphogenesis
intraciliary transport
cilium assembly
negative regulation of tyrosine phosphorylation of Stat6 protein
embryonic digit morphogenesis
negative regulation of defense response to virus
regulation of DNA binding
negative regulation of smoothened signaling pathway involved in dorsal/ventral neural tube patterning
receptor binding
protein binding
microtubule binding
tubulin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100007616.mm.1 NM_028718.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100007616.mm.1 NM_028718.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100007616.mm.1 TC0100000897.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0