This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01285498_m1
See other RGSL1 GE Assays ›
Gene Symbol:
Rgsl1
Gene Name:
regulator of G-protein signaling like 1
Gene Aliases:
4930415K13Rik, Gm202, Rgsl2
Chromosome Location:
Chr.1: 153777929 - 153847342 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01285498_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rgsl1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001243223.1 NP_001230152.1 11 - 12 2153 103
GenBank mRNA AK029630.1 - 2 - 3 260 103
AK161463.1 - 4 - 5 570 103

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100003282.mm.1

Set Membership:

Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of GTPase activity
molecular_function
GTPase activator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100026723.mm.1 NM_001243223.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100026723.mm.1 NM_001243223.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100026723.mm.1 TC0100003282.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0