This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01285138_m1
See other ZCCHC2 GE Assays ›
Gene Symbol:
Zcchc2
Gene Name:
zinc finger, CCHC domain containing 2
Gene Aliases:
9930114B20Rik, AW212015
Chromosome Location:
Chr.1: 105990406 - 106034077 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01285138_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Zcchc2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001122675.1 NP_001116147.1 9 - 10 2044 102
NM_001122676.1 NP_001116148.1 9 - 10 2044 102
GenBank mRNA AK037100.1 - 3 - 4 217 102
AK173248.1 - 9 - 10 1761 102
BC035528.1 - 6 - 7 534 102
BC082306.1 - 5 - 6 452 102

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100001016.mm.1

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function
nucleic acid binding
zinc ion binding
phosphatidylinositol binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100008417.mm.1 NM_001122675.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100008417.mm.1 NM_001122675.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100008417.mm.1 NM_001122676.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100008417.mm.1 NM_001122676.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100008417.mm.1 TC0100001016.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0