This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01284330_m1
See other CANX GE Assays ›
Gene Symbol:
Canx
Gene Name:
calnexin
Gene Aliases:
1110069N15Rik, AI988026, Cnx, D11Ertd153e
Chromosome Location:
Chr.11: 50293957 - 50325673 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01284330_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Canx
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001110499.1 NP_001103969.1 2 - 3 355 137
NM_001110500.1 NP_001103970.1 2 - 3 323 137
NM_007597.3 NP_031623.1 2 - 3 359 137
GenBank mRNA AK028279.1 - 2 - 3 286 137
AK084175.1 - 2 - 3 299 137
AK159298.1 - 2 - 3 343 137
AK165508.1 - 2 - 3 258 137
AK166649.1 - 2 - 3 336 137
BC012408.1 - 2 - 3 277 3497769 137
BC040244.1 - 2 - 3 286 2582119 137
L18888.1 - 1 - 2 171 137
L23865.1 - 1 - 2 111 137

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100002670.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Canx

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

protein folding
synaptic vesicle endocytosis
chaperone-mediated protein folding
clathrin-mediated endocytosis
glycoprotein binding
calcium ion binding
protein binding
carbohydrate binding
apolipoprotein binding
ionotropic glutamate receptor binding
poly(A) RNA binding
metal ion binding
unfolded protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100024390.mm.1 NM_001110499.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100024390.mm.1 NM_001110499.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100024390.mm.1 NM_001110500.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100024390.mm.1 NM_001110500.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100024390.mm.1 NM_007597.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100024390.mm.1 NM_007597.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100024390.mm.1 TC1100002670.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100024384.mm.1 NM_001110499.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100024384.mm.1 NM_001110499.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100024384.mm.1 NM_001110500.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100024384.mm.1 NM_001110500.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100024384.mm.1 NM_007597.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100024384.mm.1 NM_007597.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100024384.mm.1 TC1100002670.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100024387.mm.1 NM_001110499.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100024387.mm.1 NM_001110499.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100024387.mm.1 NM_007597.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100024387.mm.1 NM_007597.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100024387.mm.1 TC1100002670.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100024388.mm.1 NM_001110499.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100024388.mm.1 NM_001110499.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100024388.mm.1 NM_001110500.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100024388.mm.1 NM_001110500.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100024388.mm.1 NM_007597.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100024388.mm.1 NM_007597.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100024388.mm.1 TC1100002670.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100024385.mm.1 NM_001110499.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100024385.mm.1 NM_001110499.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100024385.mm.1 NM_001110500.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100024385.mm.1 NM_001110500.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100024385.mm.1 NM_007597.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100024385.mm.1 NM_007597.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100024385.mm.1 TC1100002670.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0