Assay Details
Gene Symbol: Fkbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013902.4 | NP_038930.1 | 6 - 7 | 636 | 112 | |
GenBank mRNA | AF110812.1 | - | 6 - 7 | 633 | 112 | |
AF135595.1 | - | 6 - 7 | 638 | 112 | ||
AK012346.1 | - | 6 - 7 | 631 | 112 | ||
AK145648.1 | - | 6 - 7 | 644 | 112 | ||
AK150802.1 | - | 6 - 7 | 646 | 112 | ||
AK168641.1 | - | 6 - 7 | 643 | 112 | ||
U62545.1 | - | 2 - 3 | 120 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28493826 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28493836 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28493825 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28493822 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28493821 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28493823 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28493819 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28493824 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28493835 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28493820 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P531175 | BC076622.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P531175 | BC029153.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P531175 | NM_177806.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P531175 | AK145648.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P531175 | AK150802.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P531175 | BC076622.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P531175 | BC029153.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P531175 | NM_177806.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P531175 | AK145648.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P531175 | AK150802.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 381755 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 376985 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 824036 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 830190 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 600556 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 948113 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 787091 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 174796 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 106849 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P155398 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P103198 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P531175 | BC076622.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P531175 | BC029153.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P531175 | NM_177806.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P531175 | AK145648.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P531175 | AK150802.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P531175 | BC076622.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P531175 | BC029153.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P531175 | NM_177806.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P531175 | AK145648.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P531175 | AK150802.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P279323 | BC029153.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P279323 | BC002122.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P279323 | AF135595.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28493816 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28493817 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28493832 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28493808 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28493797 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28493814 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28493833 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28493834 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28493835 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28493807 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28493828 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28493805 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28493799 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28493809 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28493823 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28493806 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28493821 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28493804 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28493830 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28493819 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28493826 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28493813 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28493811 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28493800 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28493827 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28493802 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07645450 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P103198 | - | - |
Whole Mouse Genome G4122A | A_52_P531175 | BC076622.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P531175 | BC029153.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P531175 | NM_177806.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P531175 | AK145648.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P531175 | AK150802.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P531175 | BC076622.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P531175 | BC029153.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P531175 | NM_177806.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P531175 | AK145648.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P531175 | AK150802.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P279323 | BC029153.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P279323 | BC002122.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P279323 | AF135595.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114562 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P103198 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10118555 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10330835 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10118560 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10330836 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10118570 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10118566 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10118557 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10330838 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10330826 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28493826 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28493824 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28493836 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28493822 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28493823 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28493825 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28493819 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28493821 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28493820 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28493835 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493776 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493775 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493823 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493786 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493783 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493695 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493780 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493686 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493772 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493757 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493824 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493685 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493841 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493873 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493770 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493750 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493855 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493814 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493843 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493845 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493807 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493846 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493782 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493837 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493833 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493883 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493828 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493805 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493792 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493762 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493849 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493787 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493796 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493751 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493753 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493761 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493689 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493778 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493866 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493804 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493819 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493683 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493821 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493881 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493693 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493802 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493691 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493878 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493806 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493853 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493756 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493809 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493799 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493773 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493765 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493862 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493832 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493864 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493759 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493816 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493764 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493875 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493789 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493784 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493800 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493835 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493830 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493817 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493760 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493852 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493684 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493811 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28493769 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28493828 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28493809 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28493797 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P09899457 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28493804 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28493780 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28493785 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28493802 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28493811 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28493772 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28493777 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28493799 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28493819 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28493782 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28493789 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28493786 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28493792 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28493787 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28493800 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28493776 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28493796 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28493817 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28493827 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28493773 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28493830 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28493793 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28493821 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28493781 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28493778 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28493816 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28493784 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10019511 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28493794 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10263508 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10323640 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28493790 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28493814 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10227120 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28493823 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10400582 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10400583 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10400588 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10400587 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10400586 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10400584 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10400585 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1416858_PM_a_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1416859_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10400581 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10396008 | - | - |
Mouse Genome 430 2.0 Array | 1416859_at | - | - |
Mouse Genome 430 2.0 Array | 1416858_a_at | - | - |
Mouse Genome 430 2.0 Array | 1416858_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1416859_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 30795_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 30795_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1200012494.mm.1 | NM_013902.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200012494.mm.1 | NM_013902.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200012494.mm.1 | TC1200001832.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1200012493.mm.1 | NM_013902.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200012493.mm.1 | NM_013902.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200012493.mm.1 | TC1200001832.mm.1 | MTA TC |
Mouse Expression Set 430 | 1416859_at | - | - |
Mouse Expression Set 430 | 1416859_PM_at | - | - |
Mouse Expression Set 430 | 1416858_PM_a_at | - | - |
Mouse Expression Set 430 | 1416858_a_at | - | - |
Murine Genome U74v2 Set | 160416_PM_at | - | - |
Murine Genome U74v2 Set | 160416_at | - | - |
Mouse Genome 430A 2.0 Array | 1416858_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1416858_PM_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1416859_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1416859_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1416858_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1416859_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1416859_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1416858_a_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 4670075 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 4670075 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004670075 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2746755 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2746755 | - | - |
mouse-6 v1.1 (Illumina) | scl42414.5.10_3-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4670075 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl42414.5.10_3-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4670075 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2746755 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2834850 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2834850 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2746755 | - | - |
