This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01283993_m1
See other ILDR2 GE Assays ›
Gene Symbol:
Ildr2
Gene Name:
immunoglobulin-like domain containing receptor 2
Gene Aliases:
2810478N18Rik, 3110063L10Rik, AI852300, D1Ertd471e, Dbsm1, ENSMUSG00000040612, Ll
Chromosome Location:
Chr.1: 166254094 - 166316832 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01283993_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ildr2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001164528.1 NP_001158000.1 4 - 5 839 61
GenBank mRNA FJ024494.1 - 785 61
FJ024495.1 - 4 - 5 785 61
FJ024496.1 - 4 - 5 785 61
FJ024500.1 - 4 - 5 785 61

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

defense/immunity protein immunoglobulin receptor superfamily

Gene Ontology Categories:

Function(s)

response to glucose
insulin secretion
cell differentiation
pancreas development
homeostasis of number of cells within a tissue

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0