This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01283855_m1
See other S100Z GE Assays ›
Gene Symbol:
S100z
Gene Name:
S100 calcium binding protein, zeta
Gene Aliases:
Gm625
Chromosome Location:
Chr.13: 95477266 - 95479379 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01283855_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: S100z
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081159.1 NP_001074628.1 1 - 2 137 113
GenBank mRNA BC147382.1 - 1 - 2 203 9056440 113
BC147383.1 - 1 - 2 203 9056460 113

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300002459.mm.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

calmodulin-related

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2013223 NM_001081159.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2013223 BC147382.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2013223 BC147383.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 382003 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 2085 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2013223 NM_001081159.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2013223 BC147382.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2013223 BC147383.1 mouse_transcriptome
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240837 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240887 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240885 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240894 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240910 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240835 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240925 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240922 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240873 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240849 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240932 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240864 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240833 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240892 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240908 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240875 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240832 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240929 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240903 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240909 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240856 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240854 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240890 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240879 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240904 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240852 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240916 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240865 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240846 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240839 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240918 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240914 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240871 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240896 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240898 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240901 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240876 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240920 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240913 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240912 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240924 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240889 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240931 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240861 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240867 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240853 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240841 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240831 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240863 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240882 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29240880 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29240898 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29240932 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10534630 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29240863 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29240924 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29240850 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29240925 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10679764 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29240870 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29240875 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29240922 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29240935 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10809249 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29240882 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29240887 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10540211 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29240868 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11064424 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29240923 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29240896 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29240903 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29240890 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29240865 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29240861 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29240871 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10534631 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29240849 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29240936 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29240904 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29240880 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29240864 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29240878 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29240848 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29240929 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11064428 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29240889 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11120701 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29240867 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10545288 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29240852 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29240916 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10754424 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29240934 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29240892 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29240885 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29240869 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11120702 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10679765 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29240919 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10578385 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29240914 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11095303 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29240859 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29240879 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29240873 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11041131 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10411224 - -
Affymetrix Mouse Gene 1.0 ST Array 10411225 - -
Affymetrix Mouse Gene 1.1 ST Array 10411223 - -
Affymetrix Mouse Clariom S Array PSR1300016432.mm.1 TC1300002459.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300016433.mm.1 NM_001081159.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300016433.mm.1 NM_001081159.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300016433.mm.1 TC1300002459.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300016434.mm.1 NM_001081159.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300016434.mm.1 NM_001081159.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300016434.mm.1 TC1300002459.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300016435.mm.1 TC1300002459.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0