This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01283227_mH
See other TIMD2 GE Assays ›
Gene Symbol:
Timd2
Gene Name:
T cell immunoglobulin and mucin domain containing 2
Gene Aliases:
AA816106, AU019358, AU019662, C78111, G9D3, TIM-2, Tim2
Chromosome Location:
Chr.11: 46668960 - 46707104 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01283227_mH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Timd2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001161355.1 NP_001154827.1 2 - 3 339 103
GenBank mRNA AK145630.1 - 2 - 3 100 103
AK163225.1 - 2 - 3 100 103
AK166877.1 - 2 - 3 93 103
CB953226.1 - 2 - 3 342 103

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK145630.1, AK163225.1, AK166877.1, NM_001161355.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0