This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01283053_m1
See other KDM6A GE Assays ›
Gene Symbol:
Kdm6a
Gene Name:
lysine (K)-specific demethylase 6A
Gene Aliases:
Utx
Chromosome Location:
Chr.X: 18162533 - 18279791 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01283053_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Kdm6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009483.1 NP_033509.1 2 - 3 571 86
GenBank mRNA AK080370.1 - 2 - 3 590 86
AK144654.1 - 2 - 3 597 86
BC053433.1 - 2 - 3 571 6516368 86

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00000190.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Kdm6a

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

in utero embryonic development
neural tube closure
heart morphogenesis
respiratory system process
heart development
regulation of gene expression
chromatin modification
neural tube development
somite rostral/caudal axis specification
multicellular organism growth
mesodermal cell differentiation
embryonic organ development
notochord morphogenesis
oxidation-reduction process
canonical Wnt signaling pathway
histone H3-K27 demethylation
cardiovascular system development
sulfonate dioxygenase activity
RNA polymerase II core promoter proximal region sequence-specific DNA binding
chromatin binding
protein binding
oxidoreductase activity
2,4-dichlorophenoxyacetate alpha-ketoglutarate dioxygenase activity
procollagen-proline dioxygenase activity
hypophosphite dioxygenase activity
identical protein binding
DNA-N1-methyladenine dioxygenase activity
metal ion binding
dioxygenase activity
C-19 gibberellin 2-beta-dioxygenase activity
C-20 gibberellin 2-beta-dioxygenase activity
histone demethylase activity (H3-K27 specific)

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00001180.mm.1 NM_009483.2 Refseq
Affymetrix Mouse Clariom S Array PSR0X00001180.mm.1 TC0X00000190.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00001178.mm.1 NM_009483.2 Refseq
Affymetrix Mouse Clariom S Array PSR0X00001178.mm.1 TC0X00000190.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0