This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01282696_g1
See other PAPD4 GE Assays ›
Gene Symbol:
Papd4
Gene Name:
PAP associated domain containing 4
Gene Aliases:
8030446C20Rik, AI649009, GLD-2, Gld2
Chromosome Location:
Chr.13: 93146275 - 93192970 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01282696_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Papd4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_133905.2 NP_598666.1 9 - 10 1124 123
GenBank mRNA AK033141.1 - 6 - 7 459 123
AK146086.1 - 9 - 10 1042 123
AM419011.1 - 8 - 9 885 123
BC016629.1 - 9 - 10 1124 3595512 123

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Papd4

Panther Classification:

Molecular Function -

nucleotidyltransferase transferase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

hematopoietic progenitor cell differentiation
mRNA processing
RNA polyadenylation
histone mRNA catabolic process
nucleotide binding
polynucleotide adenylyltransferase activity
protein binding
ATP binding
transferase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0