This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01282375_m1
See other DMGDH GE Assays ›
Gene Symbol:
Dmgdh
Gene Name:
dimethylglycine dehydrogenase precursor
Gene Aliases:
1200014D15Rik, AI787269
Chromosome Location:
Chr.13: 93674436 - 93752823 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01282375_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dmgdh
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028772.3 NP_083048.1 9 - 10 1510 62
GenBank mRNA AK004755.1 - 9 - 10 1511 62
BC024126.1 - 5 - 6 793 62
BC089599.1 - 9 - 10 1513 30118948 62

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300001051.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dmgdh

Panther Classification:

Molecular Function -

dehydrogenase methyltransferase

Gene Ontology Categories:

Function(s) Process(es)

glycine metabolic process
tetrahydrofolate interconversion
oxidation-reduction process
folic acid binding
oxidoreductase activity
poly(A) RNA binding
dimethylglycine dehydrogenase activity
flavin adenine dinucleotide binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300006709.mm.1 NM_028772.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006709.mm.1 NM_028772.3 Refseq
Affymetrix Mouse Clariom S Array PSR1300006709.mm.1 TC1300001051.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300006708.mm.1 NM_028772.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006708.mm.1 NM_028772.3 Refseq
Affymetrix Mouse Clariom S Array PSR1300006708.mm.1 TC1300001051.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0