This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01282290_m1
See other ATG10 GE Assays ›
Gene Symbol:
Atg10
Gene Name:
autophagy related 10
Gene Aliases:
5330424L23Rik, 5430428K15Rik, AI852123, APG10, Agp10, Apg10l, Apg10p, Atg10l
Chromosome Location:
Chr.13: 90935348 - 91224792 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01282290_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atg10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025770.3 NP_080046.3 2 - 3 167 60
GenBank mRNA AB079383.1 - 1 - 2 123 60
AB091691.1 - 1 - 2 132 60
AK009076.1 - 2 - 3 151 60
AK077333.1 - 2 - 3 140 60
BC023455.1 - 1 - 2 131 5249268 60

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300002407.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Atg10

Panther Classification:

Molecular Function -

ubiquitin-protein ligase protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

protein lipidation
transport
autophagy
protein transport
positive regulation of protein modification process
protein modification by small protein conjugation
autophagy in response to ER overload
protein binding
UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
ligase activity
ribosomal S6-glutamic acid ligase activity
Atg12 transferase activity
coenzyme F420-0 gamma-glutamyl ligase activity
coenzyme F420-2 alpha-glutamyl ligase activity
protein-glycine ligase activity
protein-glycine ligase activity, initiating
protein-glycine ligase activity, elongating
tubulin-glycine ligase activity
protein-glutamic acid ligase activity
tubulin-glutamic acid ligase activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300015991.mm.1 NM_025770.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300015991.mm.1 NM_025770.3 Refseq
Affymetrix Mouse Clariom S Array PSR1300015991.mm.1 TC1300002407.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300015992.mm.1 NM_025770.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300015992.mm.1 NM_025770.3 Refseq
Affymetrix Mouse Clariom S Array PSR1300015992.mm.1 TC1300002407.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 913891 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0