This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01279778_m1
See other PLA2G4D GE Assays ›
Gene Symbol:
Pla2g4d
Gene Name:
phospholipase A2, group IVD
Gene Aliases:
2610311B01Rik, Pla2delta, cPLA2delta
Chromosome Location:
Chr.2: 120265866 - 120294260 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01279778_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pla2g4d
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001024137.1 NP_001019308.1 12 - 13 1153 69
GenBank mRNA AB195276.1 - 12 - 13 1153 69
AK145985.1 - 12 - 13 1154 69
AK167492.1 - 12 - 13 1121 69
BC113160.1 - 12 - 13 1150 40060148 69
BC113770.1 - 12 - 13 1150 40060146 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200004456.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pla2g4d

Panther Classification:

Molecular Function -

phospholipase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
metabolic process
phospholipid catabolic process
lipid catabolic process
phospholipase activity
phospholipase A2 activity
hydrolase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200037370.mm.1 NM_001024137.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200037370.mm.1 NM_001024137.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200037370.mm.1 TC0200004456.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200037375.mm.1 NM_001024137.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200037375.mm.1 NM_001024137.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200037375.mm.1 TC0200004456.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200037363.mm.1 NM_001024137.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200037363.mm.1 NM_001024137.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200037363.mm.1 TC0200004456.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200037364.mm.1 NM_001024137.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200037364.mm.1 NM_001024137.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200037364.mm.1 TC0200004456.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200037367.mm.1 NM_001024137.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200037367.mm.1 NM_001024137.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200037367.mm.1 TC0200004456.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200037373.mm.1 NM_001024137.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200037373.mm.1 NM_001024137.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200037373.mm.1 TC0200004456.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200037365.mm.1 NM_001024137.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200037365.mm.1 NM_001024137.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200037365.mm.1 TC0200004456.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200037371.mm.1 NM_001024137.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200037371.mm.1 NM_001024137.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200037371.mm.1 TC0200004456.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200037372.mm.1 NM_001024137.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200037372.mm.1 NM_001024137.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200037372.mm.1 TC0200004456.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200037376.mm.1 NM_001024137.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200037376.mm.1 NM_001024137.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200037376.mm.1 TC0200004456.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200037377.mm.1 NM_001024137.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200037377.mm.1 NM_001024137.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200037377.mm.1 TC0200004456.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200037366.mm.1 NM_001024137.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200037366.mm.1 NM_001024137.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200037366.mm.1 TC0200004456.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200037368.mm.1 NM_001024137.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200037368.mm.1 NM_001024137.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200037368.mm.1 TC0200004456.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200037369.mm.1 NM_001024137.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200037369.mm.1 NM_001024137.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200037369.mm.1 TC0200004456.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200037374.mm.1 NM_001024137.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200037374.mm.1 NM_001024137.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200037374.mm.1 TC0200004456.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0