This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01279376_m1
See other SBF2 GE Assays ›
Gene Symbol:
Sbf2
Gene Name:
SET binding factor 2
Gene Aliases:
4833411B01Rik, AA414977, AI317167, B430219L04Rik, Mtmr13, mMTMH1
Chromosome Location:
Chr.7: 110308011 - 110614969 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01279376_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sbf2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_177324.2 NP_796298.2 25 - 26 3407 81
GenBank mRNA BC015069.1 - 2 - 3 198 81
CO433861.1 - 3 - 4 438 81

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700004095.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sbf2

Panther Classification:

Molecular Function -

phosphatase

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of GTPase activity
regulation of catalytic activity
protein tetramerization
Rab guanyl-nucleotide exchange factor activity
phosphatase regulator activity
phosphatase binding
phosphatidylinositol binding
protein homodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700033884.mm.1 NM_177324.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700033884.mm.1 NM_177324.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700033884.mm.1 TC0700004095.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700033886.mm.1 NM_177324.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700033886.mm.1 NM_177324.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700033886.mm.1 TC0700004095.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700033885.mm.1 NM_177324.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700033885.mm.1 NM_177324.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700033885.mm.1 TC0700004095.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0