This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01279329_g1
See other ANXA7 GE Assays ›
Gene Symbol:
Anxa7
Gene Name:
annexin A7
Gene Aliases:
AI265384, AI316497, Anx7, synexin
Chromosome Location:
Chr.14: 20455261 - 20480133 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01279329_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Anxa7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001110794.1 NP_001104264.1 4 - 5 608 70
NM_009674.3 NP_033804.2 4 - 5 723 70
GenBank mRNA AK032013.1 - 4 - 5 499 70
AK077579.1 - 4 - 5 416 70
AK134589.1 - 4 - 5 427 70
AK149654.1 - 4 - 5 415 70
AK151438.1 - 4 - 5 417 70
AK161613.1 - 4 - 5 417 70
AK167588.1 - 4 - 5 391 70
AK169199.1 - 4 - 5 390 70
AK178561.1 - 1 - 2 41 70
BC008997.1 - 4 - 5 407 3157926 70
L13129.1 - 4 - 5 410 70

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Anxa7

Panther Classification:

Molecular Function -

calcium-binding protein

Gene Ontology Categories:

Function(s) Process(es)

cellular calcium ion homeostasis
autophagy
hemostasis
cell proliferation
regulation of cell shape
response to salt stress
cellular water homeostasis
negative regulation of gene expression
response to organic cyclic compound
response to calcium ion
membrane fusion
integrin binding
calcium ion binding
protein binding
calcium-dependent phospholipid binding
poly(A) RNA binding
calcium-dependent protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400011483.mm.1 NM_001110794.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400011483.mm.1 NM_001110794.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400011483.mm.1 NM_009674.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400011483.mm.1 NM_009674.3 Refseq
Affymetrix Mouse Clariom S Array PSR1400011483.mm.1 TC1400001537.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0