This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01279326_m1
See other ECD GE Assays ›
Gene Symbol:
Ecd
Gene Name:
ecdysoneless homolog (Drosophila)
Gene Aliases:
5730461K03Rik, Hsgt1
Chromosome Location:
Chr.14: 20319859 - 20348121 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01279326_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ecd
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027475.3 NP_081751.1 5 - 6 923 97
GenBank mRNA AK017681.1 - 3 - 4 347 97
AK028664.1 - 5 - 6 887 97
AK043261.1 - 5 - 6 772 97
BC013470.1 - 5 - 6 872 4220504 97

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400001531.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ecd

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
cell proliferation
regulation of G1/S transition of mitotic cell cycle
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400011394.mm.1 NM_027475.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400011394.mm.1 NM_027475.3 Refseq
Affymetrix Mouse Clariom S Array PSR1400011394.mm.1 TC1400001531.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0