This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01279263_m1
See other ADGRF4 GE Assays ›
Gene Symbol:
Adgrf4
Gene Name:
adhesion G protein-coupled receptor F4
Gene Aliases:
4632435A09Rik, AV239010, Gpr115
Chromosome Location:
Chr.17: 42656887 - 42692284 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01279263_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Adgrf4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001289499.1 NP_001276428.1 9 - 10 2273 68
NM_001289500.1 NP_001276429.1 9 - 10 2260 68
NM_001289501.1 NP_001276430.1 9 - 10 2304 68
NM_030067.2 NP_084343.1 9 - 10 2347 68
GenBank mRNA AK019508.1 - 9 - 10 2350 68
BC140980.1 - 9 - 10 2213 9055592 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
signal transducer activity
transmembrane signaling receptor activity
G-protein coupled receptor activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1987146 AK019508.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1987146 NM_030067.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 246154 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 20944 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 726774 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 123726 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 749865 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 951132 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 872066 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 838207 AK140533.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 838207 AK142198.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 838207 NM_001039373.4 refseq_mrna_mouse_NM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 838207 NM_010839.5 refseq_mrna_mouse_NM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 838207 U32332.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 838207 Z35294.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 913884 AF125570.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 913884 AF125571.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 913884 AF125572.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 913884 AK007663.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 913884 BC028505.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 913884 NM_011332.2 refseq_mrna_mouse_NM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 126870 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 526292 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 678769 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 659187 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 725144 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 335138 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 712975 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 211764 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 216691 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 15435 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 725373 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 766746 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 147459 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 179271 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 46881 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 157870 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P142480 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P179528 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1987146 AK019508.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1987146 NM_030067.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P179528 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08780385 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08780435 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08780452 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P142480 - -
Whole Mouse Genome G4122A A_51_P304977 AK019508.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P304977 BC089564.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P304977 NM_030067.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P142480 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P179528 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185612 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185604 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185597 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13297934 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185541 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185595 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185571 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185617 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185596 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185555 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185428 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185614 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185432 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13306514 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185423 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185536 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13133096 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185545 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185544 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13157681 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185603 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185551 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185581 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185424 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185537 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13430238 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185534 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13297925 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185606 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13549983 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13398328 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185562 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185574 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185549 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185538 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185425 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13182414 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185602 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185567 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185546 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185554 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185585 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185594 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185570 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185599 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185540 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13398324 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185577 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185568 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185618 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185605 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185557 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185558 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185593 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13157676 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185565 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13082521 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185563 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185610 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185582 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13207780 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185587 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31185430 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13082524 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10450984 - -
Affymetrix Mouse Gene 1.0 ST Array 10450986 - -
Affymetrix Mouse Gene 1.0 ST Array 10450983 - -
Affymetrix Mouse Gene 1.0 ST Array 10450993 - -
Affymetrix Mouse Gene 1.0 ST Array 10450988 - -
Affymetrix Mouse Gene 1.0 ST Array 10450985 - -
Affymetrix Mouse Gene 1.0 ST Array 10450991 - -
Affymetrix Mouse Gene 1.0 ST Array 10450987 - -
Affymetrix Mouse Gene 1.0 ST Array 10450989 - -
Affymetrix Mouse Gene 1.0 ST Array 10450990 - -
Affymetrix HT MG-430 PM Array Plate 1429460_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10450982 - -
Mouse Genome 430 2.0 Array 1429460_at - -
Mouse Genome 430 2.0 Array 1429460_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 78249_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 78249_at - -
Mouse Expression Set 430 1429460_PM_at - -
Mouse Expression Set 430 1429460_at - -
Affymetrix HT Mouse Genome 430B Array 1429460_at - -
Affymetrix HT Mouse Genome 430B Array 1429460_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 580113 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 2360341 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0