This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01279260_m1
See other ADGRF4 GE Assays ›
Gene Symbol:
Adgrf4
Gene Name:
adhesion G protein-coupled receptor F4
Gene Aliases:
4632435A09Rik, AV239010, Gpr115
Chromosome Location:
Chr.17: 42656887 - 42692284 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01279260_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Adgrf4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001289499.1 NP_001276428.1 6 - 7 2113 120
NM_001289500.1 NP_001276429.1 6 - 7 2100 120
NM_001289501.1 NP_001276430.1 6 - 7 2144 120
NM_030067.2 NP_084343.1 6 - 7 2187 120
GenBank mRNA AK019508.1 - 6 - 7 2190 120
BC140980.1 - 6 - 7 2053 9055592 120

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700002114.mm.1

Set Membership:

Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
signal transducer activity
transmembrane signaling receptor activity
G-protein coupled receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700019790.mm.1 NM_001289499.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700019790.mm.1 NM_001289500.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700019790.mm.1 NM_001289501.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700019790.mm.1 NM_030067.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700019790.mm.1 TC1700002114.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0