Assay Details
Gene Symbol: Cracr2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025103.2 | NP_001020274.2 | 1 - 2 | 95 | 77 | |
GenBank mRNA | BC048916.1 | - | 1 - 2 | 95 | 77 | |
BC098333.1 | - | 1 - 2 | 56 | 5256663 | 77 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC098333.1, NM_001025103.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25588978 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25588982 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25588977 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25589002 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25588981 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25589000 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25589003 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P552879 | BC098333.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P552879 | NM_001025103.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P552879 | BC026645.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P552879 | BC034124.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P552879 | BC048916.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P552879 | BC098333.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P552879 | NM_001025103.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P552879 | BC026645.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P552879 | BC034124.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P552879 | BC048916.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 7113 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 545999 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 751903 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 50596 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 861731 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 59739 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P114840 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2367843 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P552879 | BC098333.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P552879 | NM_001025103.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P552879 | BC026645.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P552879 | BC034124.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P552879 | BC048916.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P552879 | BC098333.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P552879 | NM_001025103.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P552879 | BC026645.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P552879 | BC034124.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P552879 | BC048916.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588984 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588983 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25589011 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588988 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588994 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588991 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25589009 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25589005 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25589004 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588963 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588977 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588971 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25589012 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588997 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25589006 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25589016 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25589017 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588996 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588978 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588987 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25589001 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588992 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588972 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25589015 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588990 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588970 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588965 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588974 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588975 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25589003 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588979 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25589014 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588981 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25589000 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588968 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588986 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588967 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25589007 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06396718 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P114840 | - | - |
Whole Mouse Genome G4122A | A_52_P552879 | BC098333.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P552879 | NM_001025103.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P552879 | BC026645.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P552879 | BC034124.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P552879 | BC048916.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P552879 | BC098333.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P552879 | NM_001025103.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P552879 | BC026645.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P552879 | BC034124.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P552879 | BC048916.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117240 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P114840 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06112523 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06112458 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06112466 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06112469 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06112530 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06112452 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06112515 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25588981 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25589000 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25589003 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25588977 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25589002 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25588982 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25588978 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25589090 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25589009 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25589037 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25589000 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588988 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25589070 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588914 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588983 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25589022 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25589001 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588928 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588929 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588990 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588932 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588981 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588910 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25589014 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588979 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25589060 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25589084 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25589006 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588901 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25589028 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588968 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588922 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588937 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588977 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25589057 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588992 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588892 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588920 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588991 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25589007 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588915 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588917 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25589012 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25589035 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25589064 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25589040 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588996 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588934 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588904 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25589086 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588974 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588902 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25589023 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588994 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25589092 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25589073 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25589025 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588997 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25589041 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588975 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588926 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588903 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588899 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588891 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25589026 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25589047 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588972 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25589091 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588911 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25589019 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588888 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25589021 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25589024 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25589056 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588986 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25589049 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25589075 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588984 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588931 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588889 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25589063 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25589044 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588971 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25589020 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25589016 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25589017 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25589082 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25589050 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588965 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25589088 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25589058 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25589042 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25589079 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25589052 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588906 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588919 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25589003 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588897 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588924 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25589077 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06422451 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25588986 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25588994 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25589006 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25589003 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25588971 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25588975 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25589012 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25588979 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25588972 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25589015 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25588965 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25589007 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06539135 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25589016 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06539155 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06112523 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25588996 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25588988 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25588974 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25588967 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25589000 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06539152 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25588970 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25589017 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06112468 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06408319 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06422453 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06422455 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25589005 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25589001 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06408312 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25588990 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10558942 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10558946 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10558945 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10558947 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10558943 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10558938 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10558940 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10558941 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10558939 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10558944 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10558937 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1427367_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10558936 | - | - |
Mouse Genome 430 2.0 Array | 1427367_at | - | - |
Mouse Genome 430 2.0 Array | 1427367_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 213573_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 213573_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0700018268.mm.1 | NM_001025103.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700018268.mm.1 | NM_001025103.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700018268.mm.1 | TC0700002054.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700018273.mm.1 | NM_001025103.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700018273.mm.1 | NM_001025103.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700018273.mm.1 | TC0700002054.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700018267.mm.1 | NM_001025103.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700018267.mm.1 | NM_001025103.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700018267.mm.1 | TC0700002054.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700018274.mm.1 | NM_001025103.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700018274.mm.1 | NM_001025103.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700018274.mm.1 | TC0700002054.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700018272.mm.1 | NM_001025103.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700018272.mm.1 | NM_001025103.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700018272.mm.1 | TC0700002054.mm.1 | MTA TC |
Mouse Expression Set 430 | 1427367_at | - | - |
Mouse Expression Set 430 | 1427367_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1427367_at | - | - |
Mouse Genome 430A 2.0 Array | 1427367_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1427367_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1427367_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 659670 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.1 expression beadchip | ILMN_1238203 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2720663 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2913222 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2913222 | - | - |
