This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01278697_m1
See other EIF3C GE Assays ›
Gene Symbol:
Eif3c
Gene Name:
eukaryotic translation initiation factor 3, subunit C
Gene Aliases:
110kDa, 3230401O13Rik, Eif3s8, NIPIL(A3), NipilA3, Xs, Xsl
Chromosome Location:
Chr.7: 126546911 - 126566366 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01278697_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eif3c
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_146200.1 NP_666312.1 10 - 11 1092 68
GenBank mRNA AK088076.1 - 10 - 11 1135 68
AK160078.1 - 10 - 11 1112 68
AK168160.1 - 10 - 11 1096 68
AK168174.1 - 10 - 11 1096 68
AK197270.1 - 2 - 3 246 68
AK207372.1 - 2 - 3 146 68
BC024855.1 - 10 - 11 1092 5359060 68
BC025032.1 - 10 - 11 1113 4947435 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700004282.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eif3c

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

translation
translational initiation
RNA binding
translation initiation factor activity
translation initiation factor binding
ribosome binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700035749.mm.1 NM_146200.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700035749.mm.1 NM_146200.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700035749.mm.1 TC0700004282.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700035748.mm.1 NM_146200.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700035748.mm.1 NM_146200.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700035748.mm.1 TC0700004282.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0