This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01278560_m1
See other HIPK3 GE Assays ›
Gene Symbol:
Hipk3
Gene Name:
homeodomain interacting protein kinase 3
Gene Aliases:
DYRK6, FIST3, PKY, mir-1902
Chromosome Location:
Chr.2: 104426481 - 104494489 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01278560_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hipk3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001145824.1 NP_001139296.1 6 - 7 2160 111
NM_010434.2 NP_034564.2 6 - 7 2160 111
GenBank mRNA AF077660.1 - 6 - 7 1914 111
AF170305.1 - 5 - 6 1624 111
AF305238.1 - 5 - 6 1622 111
AK140130.1 - 6 - 7 2263 111
BC131663.1 - 6 - 7 1792 111
BC131664.1 - 6 - 7 1791 40110751 111

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200004241.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hipk3

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
protein phosphorylation
apoptotic process
mRNA transcription
phosphorylation
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
negative regulation of apoptotic process
positive regulation of DNA binding
negative regulation of JUN kinase activity
nucleotide binding
DNA binding
transcription corepressor activity
protein kinase activity
protein serine/threonine kinase activity
protein binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200035720.mm.1 NM_001145824.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200035720.mm.1 NM_001145824.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200035720.mm.1 NM_010434.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200035720.mm.1 NM_010434.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200035720.mm.1 TC0200004241.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0