This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01278507_g1
See other NR4A2 GE Assays ›
Gene Symbol:
Nr4a2
Gene Name:
nuclear receptor subfamily 4, group A, member 2
Gene Aliases:
HZF-3, NOT, Nurr1, RNR-1, TINOR, TINUR
Chromosome Location:
Chr.2: 57106830 - 57124003 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01278507_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nr4a2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001139509.1 NP_001132981.1 6 - 7 1662 105
NM_013613.2 NP_038641.1 6 - 7 1808 105
GenBank mRNA AK087495.1 - 7 - 8 1686 105
AK158607.1 - 6 - 7 1809 105
BC137715.1 - 6 - 7 2120 8860734 105
S53744.1 - 6 - 7 1699 105

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200003560.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Nuclear Receptor Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nr4a2

Panther Classification:

Molecular Function -

C4 zinc finger nuclear receptor

Gene Ontology Categories:

Function(s) Process(es)

response to hypoxia
neuron migration
response to amphetamine
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
nervous system development
adult locomotory behavior
post-embryonic development
response to inorganic substance
gene expression
regulation of gene expression
obsolete death
response to insecticide
central nervous system projection neuron axonogenesis
central nervous system neuron differentiation
habenula development
neuron differentiation
intracellular receptor signaling pathway
cellular response to extracellular stimulus
cellular response to oxidative stress
regulation of dopamine metabolic process
dopamine biosynthetic process
dopamine metabolic process
neuron maturation
positive regulation of catalytic activity
steroid hormone mediated signaling pathway
negative regulation of neuron apoptotic process
regulation of respiratory gaseous exchange
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
general adaptation syndrome
dopaminergic neuron differentiation
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II core promoter sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
steroid hormone receptor activity
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
protein binding
zinc ion binding
sequence-specific DNA binding
metal ion binding
protein heterodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0