This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01278252_m1
See other SULT1C1 GE Assays ›
Gene Symbol:
Sult1c1
Gene Name:
sulfotransferase family, cytosolic, 1C, member 1
Gene Aliases:
(PST)G, P-SULT, ST1C1, Stp2, Sult1a2, mOLFST
Chromosome Location:
Chr.17: 53961615 - 53990631 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01278252_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sult1c1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_018751.2 NP_061221.2 2 - 3 309 108
GenBank mRNA AF033653.1 - 2 - 3 320 108
BC045149.1 - 2 - 3 309 6308175 108

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700002292.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sult1c1

Panther Classification:

Molecular Function -

transferase

Gene Ontology Categories:

Function(s) Process(es)

sulfur compound metabolic process
metabolic process
N-acetylglucosamine 6-O-sulfotransferase activity
N-acetylgalactosamine 4-O-sulfotransferase activity
alcohol sulfotransferase activity
aryl sulfotransferase activity
heparan sulfate 2-O-sulfotransferase activity
sulfotransferase activity
HNK-1 sulfotransferase activity
transferase activity
heparan sulfate 6-O-sulfotransferase activity
trans-9R,10R-dihydrodiolphenanthrene sulfotransferase activity
1-phenanthrol sulfotransferase activity
3-phenanthrol sulfotransferase activity
4-phenanthrol sulfotransferase activity
trans-3,4-dihydrodiolphenanthrene sulfotransferase activity
9-phenanthrol sulfotransferase activity
2-phenanthrol sulfotransferase activity
phenanthrol sulfotransferase activity
1-hydroxypyrene sulfotransferase activity
galactose 3-O-sulfotransferase activity
proteoglycan sulfotransferase activity
cholesterol sulfotransferase activity
hydroxyjasmonate sulfotransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700021435.mm.1 NM_018751.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700021435.mm.1 NM_018751.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700021435.mm.1 TC1700002292.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700021437.mm.1 NM_018751.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700021437.mm.1 NM_018751.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700021437.mm.1 TC1700002292.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700021436.mm.1 NM_018751.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700021436.mm.1 NM_018751.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700021436.mm.1 TC1700002292.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header