This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01277984_g1
See other SLC25A12 GE Assays ›
Gene Symbol:
Slc25a12
Gene Name:
solute carrier family 25 (mitochondrial carrier, Aralar), member 12
Gene Aliases:
2610002D09Rik, AI839531, B230107K20Rik, BB129864
Chromosome Location:
Chr.2: 71274270 - 71376694 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01277984_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc25a12
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172436.3 NP_766024.1 12 - 13 1253 108
GenBank mRNA AK019150.1 - 12 - 13 1256 108
AK030595.1 - 12 - 13 1283 108
AK083156.1 - 12 - 13 1254 108
AK144958.1 - 12 - 13 1287 108
AK147948.1 - 12 - 13 1269 108
BC013845.1 - 12 - 13 1232 108
BC024952.1 - 12 - 13 1446 108
BC060505.1 - 12 - 13 1240 6311836 108

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200003720.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc25a12

Panther Classification:

Molecular Function -

transporter secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
aspartate transport
L-glutamate transport
malate-aspartate shuttle
response to calcium ion
transmembrane transport
L-glutamate transmembrane transport
L-aspartate transmembrane transport
L-glutamate transmembrane transporter activity
calcium ion binding
L-aspartate transmembrane transporter activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200031856.mm.1 NM_172436.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200031856.mm.1 NM_172436.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200031856.mm.1 TC0200003720.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0