Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200038585.mm.1 |
NM_199022.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200038585.mm.1 |
NM_199022.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200038585.mm.1 |
TC0200004529.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200038583.mm.1 |
NM_199022.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200038583.mm.1 |
NM_199022.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200038583.mm.1 |
TC0200004529.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200038584.mm.1 |
NM_199022.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200038584.mm.1 |
NM_199022.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200038584.mm.1 |
TC0200004529.mm.1 |
MTA TC |