This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01277881_g1
See other 4930473A02RIK GE Assays ›
Gene Symbol:
4930473A02Rik
Gene Name:
RIKEN cDNA 4930473A02 gene
Gene Aliases:
-
Chromosome Location:
Chr.2: 130543791 - 130563749 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01277881_g1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4930473A02Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_040348.1 8 - 9 1086 114
GenBank mRNA AK015562.1 - 8 - 9 1085 114
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BG071843.2 - 1 - 2 Chr. 2 - 130543793 - 130552235 [+] on Build GRCm38 114
GenBankmRNA AU044997.1 - 1 - 2 Chr. 2 - 130543793 - 130544515 [+] on Build GRCm38 114

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream protein tyrosine phosphatase,
receptor type,
A
Ptpra Ptpa,
Ptpalpha,
Rptpalpha,
Rptra,
Rptralpha
Anti-sense Chr. 2 - 130450252 - 130554300 on NCBI Build GRCm38
Downstream mitochondrial ribosomal protein S26 Mrps26 AI648866,
GI008,
MRP-S13,
MRP-S26,
Rpms13
19.2kb Anti-sense Chr. 2 - 130563757 - 130565394 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200004629.mm.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P284555 AK015562.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P284555 AK015562.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P284555 AK015562.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P284555 AK015562.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04692489 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04692497 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139924 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107805 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10487682 - -
Affymetrix Mouse Gene 1.0 ST Array 10487679 - -
Affymetrix Mouse Gene 1.0 ST Array 10487683 - -
Affymetrix Mouse Gene 1.0 ST Array 10487676 - -
Affymetrix Mouse Gene 1.0 ST Array 10487678 - -
Affymetrix Mouse Gene 1.0 ST Array 10487677 - -
Affymetrix Mouse Gene 1.0 ST Array 10487680 - -
Affymetrix Mouse Gene 1.0 ST Array 10487681 - -
Affymetrix Mouse Gene 1.0 ST Array 10487684 - -
Affymetrix HT MG-430 PM Array Plate 1430249_PM_at - -
Mouse Genome 430 2.0 Array 1430249_PM_at - -
Mouse Genome 430 2.0 Array 1430249_at - -
Affymetrix Mouse Clariom S Array PSR0200039585.mm.1 NR_040348.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200039585.mm.1 NR_040348.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200039585.mm.1 TC0200004629.mm.1 MTA TC
Mouse Expression Set 430 1430249_at - -
Mouse Expression Set 430 1430249_PM_at - -
Murine Genome U74v2 Set 133823_at - -
Murine Genome U74v2 Set 133823_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1430249_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1430249_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 420441 - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 106420398 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2549002 - -
Illumina Sentrix mouse-6.v1 bead array 106420398 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 106420398 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2549002 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-77574795ff-5hfml:80/100.66.128.142:80.
git-commit: 5cc8a939e1477954742eeb281a87e54f28adfb77
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header